Drukuj
Rybonukleaza bydlęca A (z trzustki
Dostawca: MP Biomedicals
0210107691 0210107680 0210107683
ICNA0210107691EA 3990 PLN
ICNA0210107691 ICNA0210107680 ICNA0210107683
Rybonukleaza bydlęca A (z trzustki
Enzymy
Dissolves readily at 2 mg/ml in analytical grade water to give a clear colorless solution.
That amount of enzyme causing the hydrolysis of RNA at a rate such that k (velocity constant) equals unity at 25 °C and pH 5,0.