Drukuj
Premiks akrylamid/bis, 29:1
Dostawca: MP Biomedicals
04800657
ICNA04800657EA 397 PLN
ICNA04800657
Premiks akrylamid/bis, 29:1
Elektroforezy, Żele Agaroza i akrylamid
Premix of ultra pure acrylamide with the cross-linking reagent N,N'-methylene-bis-acrylamide, final ratio 29:1.