Drukuj
Kolumny HPLC, Ultra PFP Propyl (USP L43)
Dostawca: Restek
9179365 9179312-700 9179315-700 9179332-700 9179335-700 9179352-700 9179355-700 9179362-700 9179365-700 9179512-700 9179515-700 9179522-700 9179525-700 9179532-700 9179535-700 9179552-700 9179555-700 9179562-700 9179565-700 9179575-700 9179311 9179312 9179315 9179331 9179332 9179335 9179351 9179352 9179355 9179361 9179362 9179511 9179512 9179515 9179521 9179522 9179525 9179531 9179532 9179535 9179551 9179552 9179555 9179561 9179562 9179565 9179571 9179572 9179575 917931E 917933E 917935E 917936E 917951E 917952E 917953E 917955E 917956E 9179572-700 917957E
REST9179365EA 0 PLN
REST9179365 REST9179312-700 REST9179315-700 REST9179332-700 REST9179335-700 REST9179352-700 REST9179355-700 REST9179362-700 REST9179365-700 REST9179512-700 REST9179515-700 REST9179522-700 REST9179525-700 REST9179532-700 REST9179535-700 REST9179552-700 REST9179555-700 REST9179562-700 REST9179565-700 REST9179575-700 REST9179311 REST9179312 REST9179315 REST9179331 REST9179332 REST9179335 REST9179351 REST9179352 REST9179355 REST9179361 REST9179362 REST9179511 REST9179512 REST9179515 REST9179521 REST9179522 REST9179525 REST9179531 REST9179532 REST9179535 REST9179551 REST9179552 REST9179555 REST9179561 REST9179562 REST9179565 REST9179571 REST9179572 REST9179575 REST917931E REST917933E REST917935E REST917936E REST917951E REST917952E REST917953E REST917955E REST917956E REST9179572-700 REST917957E
Kolumny HPLC, Ultra PFP Propyl (USP L43)
Kolumny chromatograficzne
Ultra PFP Propyl columns are with a pentafluorophenyl propyl phase and has retention and selectivity of charged bases, electronegative compounds and amine-containing compounds.
These are reliable and efficient with acidic mobile phases. The versatile column is suitable for highly aqueous mobile phases and HILIC separations.