Drukuj
Kolumny HPLC, ZORBAX® 80 Å Extend-C18
Dostawca: Agilent
770450-902 764953-902 727975-302
554-1284EA 5970 PLN
554-1284 554-1283 HEWL727975-302
Kolumny HPLC, ZORBAX® 80 Å Extend-C18
Kolumny chromatograficzne
High sensitivity for LC/MS separations of peptides.
  • High efficiency and long life at high pH (up to 11,5)
  • Unique bidentate bonding and double encapping provides high pH stability
  • Improve retention, resolution and peak shape of basic compounds
More efficiency and better peak shape than polymer-based columns.