Drukuj
Zestawy do wizualnego porównywania kolorów, cynk, MColortest™, Supelco®
Dostawca: Merck
1.14412.0001 1.14782.0002 1.14780.0001
1.14412.0001EA 789 PLN
1.14412.0001 1.14782.0002 1.14780.0001
Zestawy do wizualnego porównywania kolorów, cynk, MColortest™, Supelco®
Zestawy do testów środowiskowych
Quick tests, visual colour comparison.
  • Results in approximately five minutes
  • Easy to use
  • Application-oriented measuring ranges
  • Portable
  • A range of economical refill packs (RP) are available
MQuant® with colour card comparator
By using cuvettes with a long light path, the MQuant® (MCC) test kits are especially suited for measuring low ion concentrations (some as low as 0,005 mg/L). These test kits are especially appropriate for analysing drinking water, spring water, purified and mineral water, as well as cooling and boiler water.
MQuant® with colour disc comparator MCD
Using the comparator disc, these test kits are especially suitable for the analysis of turbid and coloured waters. The comparator disc is a practical and robust colour comparison device that provides reliable measurements, even in humid environments. Ideal for samples of medium to high concentration in applications ranging from testing bottled water to wastewater and other industrial water testing.
Dodatkowe informacje: Each test kit contains all the necessary reagents, reaction cuvettes, colour comparison chart or comparator wheel and extensive instructions for use.