Drukuj
Kombinowane elektrody pH, sensION™+
Dostawca: Hach
LZW5014T97002 LZW5011T97002 LZW5010T97002 LZW5021T97002
HACHLZW5014T97002EA 2190 PLN
HACHLZW5014T97002 662-1628 HACHLZW5010T97002 HACHLZW5021T97002
Kombinowane elektrody pH, sensION™+
Elektrody Elektrody pH
3-in-1 laboratory combination pH electrodes with built-in temperature sensor.
  • 5010T and 5011T are low maintenance electrodes for general applications
  • 5014T and 5021T are high performance electrodes for special or difficult applications
  • For use with Hach sensION+ laboratory pH meters
Dodatkowe informacje: Supplied with basic user manual. 5014T and 5021T also include 3,0 M KCl electrolyte refill solution (30 ml).