Zestawy do wizualnego oznaczania barwy, miedź, MQuant®, Supelco®
Dostawca: Merck
Quick tests for visual colour comparison.
- Results in approximately five minutes
- Easy to use
- Application-oriented measuring ranges
- Portable
- A range of economical refill packs (RP) are available
MQuant® with colour card comparator
By using cuvettes with a long light path, the MQuant® test kits are especially suited for measuring low ion concentrations (some as low as 0,005 mg/l). These test kits are especially appropriate for analysing drinking water, spring water, purified and mineral water, as well as cooling and boiler water.
RP = Refill pack
Dodatkowe informacje: Each test kit contains all the necessary reagents, reaction cuvettes, colour comparison chart and extensive instructions for use.