Drukuj
VWR®, Pakiety proszku do oznaczania molibdenu, odczynniki do fotometrów
Dostawca: VWR Chemicals
85610.620EA 854 PLN
85610.620 85611.620
VWR®, Pakiety proszku do oznaczania molibdenu, odczynniki do fotometrów
Zestawy do testów środowiskowych
VWR® Powder Pack reagents are designed to be used on laboratory and portable photometers with 16 mm cell capability. No calibration data is necessary. The pillows, individually sealed in aluminium foil packs, are unaffected by ambient conditions and contain pre-measured powder reagents for accuracy, convenience and great solubility.
  • Accurate
  • Fast and convenient by easier dissolution than tablets and long shelf life
  • No calibration necessary
  • Beveled edge prevents reagent from getting stuck in corners