Benzo[b]thiophene-3-acetic acid ≥98%
Dostawca: Thermo Fisher Scientific
|
Uwaga
|
L05855.04
L05855.09
L05855.ME
L05855.04EA
907
PLN
L05855.04
L05855.09
L05855.ME
Benzo[b]thiophene-3-acetic acid ≥98%
Purity: 98+%
CAS No.: 1131-09-5
Molecular Formula: C10H8O2S
Formula Weight: 192.24
Melting Point: 108-112°
Beilstein Reference: 143357
MDL No.: MFCD00051637
Formuła:
C₁₀H₈O₂S Temp. topnienia: 108…112 °C Temperatura przechowywania: Temperatura otoczenia |
Numer MDL:
MFCD00051637 Numer CAS: 1131-09-5 Numer WE (dawny EINECS): 214-461-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...