Teicoplanin ≥80% A2
Dostawca: Thermo Fisher Scientific
Purity: 80%
CAS No.: 61036-62-2
Storage Temperature: 2°C to 8°C
Physical Form: Powder
Appearance: White to off-white
Solubility: Sparingly soluble in water
MDL No.: MFCD05664625
Applications: Targets primarily gram positive bacteria especially those resistant to methicillin
Formuła:
C₈₉H₉₉Cl₂N₉O₃₃ Temperatura przechowywania: Lodówka |
Numer MDL:
MFCD05664625 Numer CAS: 61036-62-2 Klasyfikacja Merck : 14,9115 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...