Flecainide acetate 98%
Dostawca: Thermo Fisher Scientific
|
Uwaga
|
Purity: 98%
CAS No.: 54143-56-5
Molecular Formula: C17H20F6N2O3.C2H4O2
Formula Weight: 474.40
Storage Temperature: -30°C to -10°C
Physical Form: Powder
Appearance: White
Melting Point: 145-147°
MDL No.: MFCD00214290
Applications: Sodium channel blocker
Formuła:
C₁₇H₂₀F₆N₂O₃·C₂H₄O₂ Temp. topnienia: 145…147 °C |
Numer MDL:
MFCD00214290 Numer CAS: 54143-56-5 Numer WE (dawny EINECS): 258-997-5 Klasyfikacja Merck : 14,4098 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...