Nitrendipine
Dostawca: Thermo Fisher Scientific
|
Uwaga
|
J60841.MC
J60841.03
J60841.MCEA
1540
PLN
J60841.MC
J60841.03
Nitrendipine
Nitrendipine
CAS No.: 39562-70-4
Molecular Formula: C18H20N2O6
Formula Weight: 360.36
Physical Form: Powder
Appearance: Yellow to green
MDL No.: MFCD00082255
Applications: L-type calcium channel blocker
Formuła:
C₁₈H₂₀N₂O₆ Temperatura przechowywania: Temperatura otoczenia |
Numer MDL:
MFCD00082255 Numer CAS: 39562-70-4 Numer WE (dawny EINECS): 254-513-1 Klasyfikacja Merck : 14,6576 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...