2-(Cyclopropylmethoxy)pyridine-3-carboxaldehyde 97%
Dostawca: Thermo Fisher Scientific
|
Uwaga
|
H50091.03
H50091.06
H50091.03EA
784
PLN
H50091.03
H50091.06
2-(Cyclopropylmethoxy)pyridine-3-carboxaldehyde 97%
2-(Cyclopropylmethoxy)pyridine-3-carboxaldehyde
Purity: 97%
Molecular Formula: C10H11NO2
Formula Weight: 177.20
MDL No.: MFCD07781136