Dihydroxybis(ammonium lactato)titanium(IV) 50% (w/w) in aqueous solution
Dostawca: Thermo Fisher Scientific
|
Uwaga
|
42676.A1
42676.A7
42676.30
42676.A1EA
1820
PLN
42676.A1
42676.A7
42676.30
Dihydroxybis(ammonium lactato)titanium(IV) 50% (w/w) in aqueous solution
Dihydroxybis(ammonium lactato)titanium(IV)
CAS No.: 65104-06-5
Molecular Formula: C6H18N2O8Ti
Formula Weight: 294.10
Flash Point: 47°(118°F)
Physical Form: Liquid
Density: 1.21
Refractive Index: 1.4380
Appearance: Pale yellow
UN No.: UN1993
MDL No.: MFCD00072736
Applications: Water soluble titanium complex
Formuła:
C₆H₂₂N₂O₈Ti Gęstość: 1,21 Temp. Zapłonu: 47 °C (118 °F) Temperatura przechowywania: Temperatura otoczenia |
Numer MDL:
MFCD00072736 Numer CAS: 65104-06-5 Numer WE (dawny EINECS): 247-428-6 Jednostka: 1993 ADR: 3,III |