TCLP Standard Solution 1, Specpure®
Dostawca: Thermo Fisher Scientific
36743.AE
36743.AP
36743.AEEA
1310
PLN
36743.AE
36743.AP
TCLP Standard Solution 1, Specpure®
Standardy
Wzorce kationowe
Multi-Element Standards
UN No.: UN3264
MDL No.: MFCD02091705