Zestawy testowe, siarczyn, Spectroquant®, Supelco®
Dostawca: Merck
All Spectroquant® test kits can be used with the Prove range of spectrophotometers and Nova 60/60A instruments. Tests can be used not only on photometers and spectrophotometer from Merck, but also on photometers and spectrophotometers from other suppliers (programming details available on request). Spectroquant® cell test kits.
- Very user friendly, validated methods
- All the necessary reagents for the analysis are contained in the cells for most the test parameters
- Packaging design protects the cells from fluctuation of temperature and moisture during storage and transport
- The barcode on the cells ensures the correct method of analysis is chosen - eliminating mistakes and reducing set-up times
Spectroquant® reagent test kits
Highly stable, ready to use reagent mixtures in a user friendly design.
Free selection of appropriate cell format according to the measuring range.
Long shelf life up to 3 years.
All at room temperature.
Automatic selection of the correct method with help of AutoSelector barcode identification.
Note: The range quoted in the table below refers to the Prove and NOVA instruments. For some test kits the measuring range available for the Spectroquant® Multy and Move 100 instruments may be reduced.