Drukuj
Filtry membranowe, typ 250
Dostawca: Sartorius
25006-50-N 25006-90-G 25006-13-N 25006-25-N 25006-47-N 25007-13-N 25007-25-N 25007-47-N 25007-90-G 25007-50-N
512-7913EA 867 PLN
512-7913 SART25006-90-G 512-7116 611-0721 611-0722 512-7118 611-0723 611-0724 512-7912 SART25007-50-N
Filtry membranowe, typ 250
Filtry Filtry membranowe
Hydrophilic polyamide (nylon) membrane filters. For the filtration of alkaline solutions and organic solvents.