Drukuj
Filtry membranowe, typ 184
Dostawca: Sartorius
18406-100-G 18406-13-N 18406-25-N 18406-47-N 18406-50-N 18407-100-G 18407-13-N 18407-25-N 18407-47-N 18407-50-N 18407-142-N 18406-50-ACN
512-6516EA 898 PLN
512-6516 512-6116 512-6216 512-6316 512-6416 512-6518 512-6118 512-6218 512-6318 512-6418 SART18407-142-N SART18406-50-ACN
Filtry membranowe, typ 184
Filtry Filtry membranowe
Regenerated cellulose hydrophilic membrane filters. For filtration of organic solvents. Suitable for particle removal from aqueous solutions and solvents. Low adsorption.
* Sterile