Drukuj
Podblatowe szafki bezpieczeństwa typ 90 UTS ergo line XLT-5500 mm głębokości
Dostawca: DUPERTHAL
29-061457-401 29-061457-001 29-061459-001
DUEH29-061457-401EA 13890 PLN
DUEH29-061457-401 DUEH29-061457-001 DUEH29-061459-001
Podblatowe szafki bezpieczeństwa typ 90 UTS ergo line XLT-5500 mm głębokości
Meble Szafy bezpieczeństwa
These safety cabinets are fire resistant up to 90 minutes. These under bench cabinets are equipped with wing doors and/or drawers for the storage of hazardous substances in working spaces.
  • Easy moving, single-handed operation of the doors and drawers
  • All UTS ergo line models are fitted with a cylinder lock as a standard
  • Two door technologies are available: the classical on or the push-to open wing door technology (the easy moving safety doors are opened by applying a gentle push to the front)
Drawers or wing doors remain open in any position and containers can be inserted and removed using both hands. The drawers/doors close automatically in case of fire. All UTS models come with standard right hand door hinge, optionally available with left hand door hinge (on request).
Standard equipotential bonding saddle on the roof of the cabinet for connecting to the earthing in accordance with TRBS 2153, BGR 132 (avoiding ignition hazards). The interior fittings are conductively connected with the equipotential bonding saddle.
Constant floor extraction by the slotted base (height 35 mm). This enables harmful substances or vapours which collect at the bottom to be safely picked up and removed with the exhaust air. The exhaust connection diameter is 75 mm on the rear of the cabinet. The cut-off flaps close automatically in the case of fire.
Wing doors and carcass made from sheet steel, powder-coated. Interior fittings made from powder-coated sheet steel. Two bottom trays, two perforated sheet inserts and classical base 35 mm.
Certyfikaty: In accordance with TRbF 20 Annex L. Type testing according to DIN EN 14470-1 and DIN EN 14727 (laboratory furniture standard) by recognised test institutes (TÜV Süd, ift Rosenheim) and classification type 90 in accordance with DIN EN 14470-1, GS/CE mark.
Dodatkowe informacje: Delivery is not included, prices available upon request.