Drukuj
Avantor® ACE® C18-HL, kolumny HPLC, 10 µm
Dostawca: Avantor
ACE-331-0510EA 0 PLN
ACE-331-0510 ACE-331-0520 ACE-331-0530 ACE-331-1010 ACE-331-1020 ACE-331-1030 ACE-331-1220 ACE-331-1510 ACE-331-1520 ACE-331-1530 ACE-331-2510 ACE-331-2520 ACE-331-2530 ACE-331-2546 ACE-331-7520 ACE-331-0103GD ACE-331-0110GD ACE-331-7530 ACE-331-3020 ACE-331-3030
Avantor® ACE® C18-HL, kolumny HPLC, 10 µm
Kolumny chromatograficzne
Avantor® ACE® C18-HL columns provide great reproducibility and column lifetime. These stainless steel columns are available in a wide range of particle sizes and dimensions from capillary to preparative.
  • Ultra high purity, base deactivated silica with exceptional reproducibility
  • Ideal for a wide range of analytes differing in hydrophobicity
  • Wide range of particle sizes (3 to 15 µm) and complementary phases
  • Especially suited for preparative work due to higher carbon load
  • Method validation kits contain three different batches of the same phase - available on request
These columns are designed for a wide range of chromatographic applications, to provide great performance with acidic, basic and neutral molecules.