Drukuj
Sita testowe 100×40 mm
Dostawca: Retsch GmbH
60.108.000150 60.108.000020 60.108.000025 60.108.000032 60.108.000038 60.108.000045 60.108.000053 60.108.000063 60.108.000075 60.108.000090 60.108.000106 60.108.000125 60.108.000180 60.108.000212 60.108.000250 60.108.000300 60.108.000355 60.108.000425 60.108.000500 60.108.000600 60.108.000710 60.108.000850 60.108.001000 60.108.001180 60.108.001400 60.108.001700 60.108.002000 60.108.002360 60.108.002800 60.108.003350 60.108.004000 60.108.004750 60.108.005600 60.108.006300 60.108.006700 60.108.008000 60.108.009500 60.108.011200 60.108.012500 60.108.013200 60.108.016000 60.108.019000 60.108.022400 60.108.025000 60.108.026500 60.108.031500 60.108.037500 60.108.045000
510-5412EA 580 PLN
510-5412 510-5401 510-5402 510-5403 510-5404 510-5405 510-5406 510-5407 510-5408 510-5409 510-5410 510-5411 510-5413 510-5414 510-5415 510-5416 510-5417 510-5418 510-5419 510-5420 510-5421 510-5422 510-5423 510-5424 510-5425 510-5426 510-5427 510-5428 510-5429 510-5430 510-5431 510-5432 510-5433 510-5434 510-5435 510-5436 510-5437 510-5438 510-5439 510-5440 510-5441 510-5442 510-5443 510-5444 510-5445 510-5446 510-5447 510-5448
Sita testowe 100×40 mm
Sita Sita testowe
Sieves made from 304 stainless steel are individually laser engraved with clear and precise labelling for full traceability. They are fully compatible with Retsch's existing products and can be combined with other sieves without problems.
  • Sieve bodies are made in one piece and the absence of hollow spaces prevents cross-contamination
  • High corrosion resistance and easy to clean
  • Maximum stability and optimal sealing of the sieve stack, 15% less weight than traditional sieves
Sieve meshes, frames and labelling comply with standards. The sieves are tested five times and are delivered with test report.
Certyfikaty: An individual inspection certificate for test materials monitoring according to ASTM E 11 is also available on request.
Informacje do zamówienia: Sieves and accessories of other dimensions are available on request. Also available with perforated plate, round or square. For a full overview, please go to our website.