Drukuj
Krążki filtracyjne, seria 16, porowatość 1, VitraPOR®
Dostawca: ROBU GLASFILTER
16101 16301 16401 16501 16601 16701 16801 16901 16111 16121 16201 16251 62001 61901 61301 62601 62801 62701 62501 62401 63801 63501 63001 62901 61801 61701 61501 61401 61601 62301 62201 62101
ROBU16101EA 328 PLN
ROBU16101 ROBU16301 ROBU16401 ROBU16501 ROBU16601 ROBU16701 ROBU16801 ROBU16901 ROBU16111 ROBU16121 ROBU16201 ROBU16251 ROBU62001 ROBU61901 ROBU61301 ROBU62601 ROBU62801 ROBU62701 ROBU62501 ROBU62401 ROBU63801 ROBU63501 ROBU63001 ROBU62901 ROBU61801 ROBU61701 ROBU61501 ROBU61401 ROBU61601 ROBU62301 ROBU62201 ROBU62101
Krążki filtracyjne, seria 16, porowatość 1, VitraPOR®
Filtry Krążki filtracyjne
The unique porous structure of VitraPOR® sintered glass filters makes it flexible and versatile for many applications.
  • Very good chemical resistance
  • Very good temperature resistance
Made of borosilicate glass. The faces of filter discs from the series 16 are precisely ground down to tight tolerances and plane surfaces as may be required in specific installation circumstances.