Drukuj
Drabinka DNA, drabinka ultra niskigo zakresu II, peqGOLD
PEQL25-4110EA 485 PLN
PEQL25-4110
Drabinka DNA, drabinka ultra niskigo zakresu II, peqGOLD
Ultra Low Range DNA Ladder II for quantitative and qualitative analysis of very small dsDNA.
  • Range: 10 fragments, 25 to 700 bp
  • Two highlighted bands for easy orientation
  • Supplied with separate loading buffer or in 'Ready to Use' format