Drukuj
Głowice i miski obudowy PFA, do filtrów nabojowych, Chemlock®
Dostawca: ENTEGRIS
YY56HA20A YY5610102 YY561120A YY56HF20F_U YY5611160 YY5600010 YY5611170 YY5611180 YY5611250 YY5611253 YY5611280 YY5620170 YY5620180 YY5620250 YY5620253 YY5620280 YY56H000B YY56HB20B YY56HF24F YY56HP203 YY56HP20P YY56HP243 YY56HP24P YY5611170GF
ENTEYY56HA20AEA 0 PLN
ENTEYY56HA20A ENTEYY5610102 ENTEYY561120A ENTEYY56HF20F_U ENTEYY5611160 ENTEYY5600010 ENTEYY5611170 ENTEYY5611180 ENTEYY5611250 ENTEYY5611253 ENTEYY5611280 ENTEYY5620170 ENTEYY5620180 ENTEYY5620250 ENTEYY5620253 ENTEYY5620280 ENTEYY56H000B ENTEYY56HB20B ENTEYY56HF24F ENTEYY56HP203 ENTEYY56HP20P ENTEYY56HP243 ENTEYY56HP24P ENTEYY5611170GF
Głowice i miski obudowy PFA, do filtrów nabojowych, Chemlock®
Filtry Wkłady filtracyjne
PFA cartridge filter housing is the compact design, space-saving, enabes hassle-free cartridge changeouts (for safety and cleanliness) and reduced exposure to chemicals. It consists of lock-in design which perfectly seats the cartridge in the filter housing.
  • Smaller footprint, minimal clearance cartridge/bowl design
  • Cartridge locks into bowl with simple ¹/₄-turn twist
  • Compact head with moulded on fittings
Operating temperature (max.): 100 °C
Operating pressure (max.): 43 psi at 100 °C and 110 psi at 25 °C