Drukuj
Kombinezon, Hopen®
Dostawca: Medicom
40.001/05 40.001/04 40.001/03 40.001/02
HOPE40.001/05EA 347 PLN
HOPE40.001/05 HOPE40.001/04 HOPE40.001/03 HOPE40.001/02
Kombinezon, Hopen®
Odzież jednorazowa
These disposable overalls with hood are made of breathable PP.
  • Lightweight polypropylene is highly breathable
  • With elasticated hood, cuffs and ankles
  • With zip closure (without flap)
Ideal for use in the chemical and petrochemical industry, for food applications, cleaning and non hazardous industrial applications.
Weight: 40 g/m²