Drukuj
Anti-PSD95 Mouse Monoclonal Antibody [clone: 6G6]
Dostawca: Cayman Chemical
10011435-100 10011435-25
CAYM10011435-100EA 4030 PLN
CAYM10011435-100 CAYM10011435-25
Anti-PSD95 Mouse Monoclonal Antibody [clone: 6G6]
Przeciwciała
Type: Primary
Antigen: PSD95
Clonality: Monoclonal
Clone: 6G6
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Bovine, Mouse, Rat