Drukuj

Anti-MALT1 Mouse Monoclonal Antibody [clone: MT1/410]

Dostawca: US Biological
138567 138567-100
USBI138567EA 1970 PLN
USBI138567 USBI138567-100
Anti-MALT1 Mouse Monoclonal Antibody [clone: MT1/410]
Przeciwciała
Recommended dilution: ; Immunofluorescence: 1-2?g/ml; Western Blot: 0.5-1 ?g/ml; Immunochemistry: 0.5-1 ?g/ml; Immunoprecipitation: 1-2?g/500 ?g

Type: Primary
Antigen: MALT1
Clonality: Monoclonal
Clone: MT1/410
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human,
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR