Drukuj

Anti-Glypican 4 Rabbit Polyclonal Antibody

Dostawca: United States Biological
USBIG8235-52E-100EA 5620 PLN
USBIG8235-52E-100 USBIG8235-52E-20
Anti-Glypican 4 Rabbit Polyclonal Antibody
Przeciwciała
Recommended dilution: Western Blot: 1:1000-1:2000; Immunochemistry: 1:1000

Type: Primary
Antigen: Glypican 4
Clonality: Polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human,
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR