Drukuj

Anti-IgG (H+L) Goat Polyclonal Antibody (DyLight® 350)

Dostawca: Agrisera
AGRIAS122572EA 878 PLN
AGRIAS122572
Anti-IgG (H+L) Goat Polyclonal Antibody (DyLight® 350)
Przeciwciała
Type: Secondary
Antigen: IgG (H+L)
Clonality: Polyclonal
Clone:
Conjugation: DyLight® 350
Epitope:
Host: Goat
Isotype:
Reactivity: Bovine, Chicken, Goat, Guinea Pig, Hamster, Horse, Human, Mouse, Rabbit, Rat, Sheep
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR