Drukuj
Anti-Biotin Goat Polyclonal Antibody (DyLight® 350)
Dostawca: Agrisera
AGRIAS122481EA 1240 PLN
AGRIAS122481
Anti-Biotin Goat Polyclonal Antibody (DyLight® 350)
Przeciwciała
Type: Secondary
Antigen: Biotin
Clonality: Polyclonal
Clone:
Conjugation: DyLight® 350
Epitope:
Host: Goat
Isotype:
Reactivity:
Zobacz ceny