Drukuj

Anti-IgG (H+L) Goat Polyclonal Antibody (DyLight® 550)

Dostawca: Agrisera
AGRIAS121945EA 1100 PLN
AGRIAS121945
Anti-IgG (H+L) Goat Polyclonal Antibody (DyLight® 550)
Przeciwciała
Type: Secondary
Antigen: IgG (H+L)
Clonality: Polyclonal
Clone:
Conjugation: DyLight® 550
Epitope:
Host: Goat
Isotype:
Reactivity: Bovine, Goat, Human, Mouse, Rabbit, Rat
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR