Drukuj

Anti-IgG (H+L) Donkey Polyclonal Antibody (HRP (Horseradish Peroxidase))

Dostawca: Agrisera
AGRIAS101160EA 1240 PLN
AGRIAS101160
Anti-IgG (H+L) Donkey Polyclonal Antibody (HRP (Horseradish Peroxidase))
Przeciwciała
Type: Secondary
Antigen: IgG (H+L)
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Donkey
Isotype:
Reactivity: Bovine, Human, Mouse, Rabbit
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR