Drukuj

Anti-IgG (H+L) Chicken Polyclonal Antibody (AP (Alkaline Phosphatase))

Dostawca: Agrisera
AGRIAS101125EA 1240 PLN
AGRIAS101125
Anti-IgG (H+L) Chicken Polyclonal Antibody (AP (Alkaline Phosphatase))
Przeciwciała
Type: Secondary
Antigen: IgG (H+L)
Clonality: Polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope:
Host: Chicken
Isotype:
Reactivity: Bovine, Goat, Human, Mouse, Rabbit, Rat
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR