Drukuj

Anti-IgG (H+L) Goat Polyclonal Antibody (AP (Alkaline Phosphatase))

Dostawca: Agrisera
AGRIAS09620EA 1100 PLN
AGRIAS09620
Anti-IgG (H+L) Goat Polyclonal Antibody (AP (Alkaline Phosphatase))
Przeciwciała
Type: Secondary
Antigen: IgG (H+L)
Clonality: Polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope:
Host: Goat
Isotype:
Reactivity: Human, Mouse, Rat
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR