Drukuj
Zamknięcia, HDPE, 11 mm
Dostawca: Thermo Fisher Scientific
362830-0111 342830-0111 342830-0112 342830-0114 362830-0112 362830-0113 362830-0114 362830-0115 362830-0116 362830-0118 362830-0119 362830-1110 362830-5116 362830-5114 362830-5118 342830-0110 342830-0113 342830-0115 342830-0116 342830-0117 342830-0118 342830-0119 342830-1110 342830-5110 342830-5114 342830-5116 342830-5118 362830-0110 362830-0117 362830-5110
NALG362830-0111EA 1450 PLN
NALG362830-0111 479-0884 479-0885 479-0887 NALG362830-0112 NALG362830-0113 NALG362830-0114 NALG362830-0115 NALG362830-0116 NALG362830-0118 NALG362830-0119 NALG362830-1110 NALG362830-5116 NALG362830-5114 NALG362830-5118 NALG342830-0110 479-0886 479-0888 479-0889 479-0890 NALG342830-0118 NALG342830-0119 NALG342830-1110 NALG342830-5110 NALG342830-5114 NALG342830-5116 NALG342830-5118 NALG362830-0110 NALG362830-0117 NALG362830-5110
Zamknięcia, HDPE, 11 mm
Nakrętki Nakrętki do vialek chromatograficznych
Identify samples quickly with high profile closures with colour coders for micro packaging vials. HDPE closures are ready to use and have no O-ring to fall out or contaminate contents.
  • RNase/DNase free (only sterile closures), non-cytotoxic and non-pyrogenic
  • Permanently attached, colour coded inserts are free of toxic heavy metals
  • Inserts do not come in contact with vial contents
Recover the entire contents of Thermo Scientific™ Nalgene™ PPCO micro packaging vials with E-beam irradiation.These vials are molded from high purity, noncytotoxic, nonpyrogenic, low-metal content polypropylene copolymer resin and feature a conical bottom. Skirted vials can be centrifuged at 13,000×g.
Vials and closures (listed separately) pressure-tested together at 7,5 PSIG, USP Class VI compliant. Components provided sterile (e-beam irradiation) and ready to fill, sterile to 10 to 6 SAL.Coloured closures packaged separately, are leakproof† when used with Nalgene™ closures.
Certyfikaty: Manufactured in compliance with ISO 13485:2003 quality requirements. Micro packaging vials and closures meet the requirements of 21CFR177.1520 for food and beverage use, USP Class VI.
Sposób pakowania: Closures are bulk packed without vials, 500 per inner bag, 1000 per case.