Drukuj
Systemy oczyszczania wody, seria HLP
Dostawca: HYDROLAB
DH-0004-00 DH-0005-00 DH-0005-0S DH-0010-00 DH-0010-0S DH-0010-0P DH-0020-SP DH-0030-0P DH-0030-SP DH-0005-SP DH-0020-0P DH-0010-UV DH-0005-UV DH-0010-SP DH-0020-UV DH-0030-UV DH-0005-0P
HYDRDH-0004-00EA 4790 PLN
HYDRDH-0004-00 HYDRDH-0005-00 HYDRDH-0005-0S HYDRDH-0010-00 HYDRDH-0010-0S HYDRDH-0010-0P HYDRDH-0020-SP HYDRDH-0030-0P HYDRDH-0030-SP HYDRDH-0005-SP 171-0843 HYDRDH-0010-UV HYDRDH-0005-UV HYDRDH-0010-SP HYDRDH-0020-UV HYDRDH-0030-UV HYDRDH-0005-0P
Systemy oczyszczania wody, seria HLP
Systemy oczyszczania wody Systemy czystej i ultra czystej wody
HLP demineralisers are fed by tap water and deliver purified water (Type II and III) for instrumental and analytical purposes. Type I water can be supplied when the device is equipped with an optional UV lamp and a microfiltration capsule (0,2 μm).
  • System is equipped with a 10 L pressure storage tank
  • Microprocessor control and measurement system
  • Conductometer measures purified water pressure (in µS/cm or MOhm)
  • Alarm informs about the necessity to replace the filters, exchange resins, UV lamp, or microfiltration capsule
  • RS232 connection for the communication with a PC
  • Safety function: Pump shuts down when the feed water pressure is too low or when the tank is full