Drukuj
Aplikator filmu, Baker
Dostawca: Industrial Physics
VF1500 VF2147 VF2146 VF2145 VF1510 VF1515 VF1502 VF1501 VF1520 VF1521 VF1522 VF1523 VF1525 VF1526 VF1527 VF1528 VF1560 VF1531
SIMEVF1500EA 2600 PLN
SIMEVF1500 SIMEVF2147 SIMEVF2146 SIMEVF2145 SIMEVF1510 SIMEVF1515 SIMEVF1502 SIMEVF1501 SIMEVF1520 SIMEVF1521 SIMEVF1522 SIMEVF1523 SIMEVF1525 SIMEVF1526 SIMEVF1527 SIMEVF1528 SIMEVF1560 SIMEVF1531
Aplikator filmu, Baker
Aplikator filmu uszczelniającego Aplikatory farb / tuszów
Cylindrical film applicator with four application sides for applying paint films of four different predefined thicknesses in film width 60 or 80 mm. Suitable for applying a host of different products onto flat and relatively solid substrates. Made from high grade stainless steel, the film applicator will not be affected by acid or base elements.
Dodatkowe informacje: Supplied in a plastic case with calibration certificate.