Drukuj
Wodoodporny termometr
Dostawca: CERTOCLAV
CERT1300149_BEA 0 PLN
CERT1300149_B
Wodoodporny termometr
Waterproof thermometer for accurate measurements in labs, quality management, and production processes. Applicable in food (HACCP), medical and pharmaceutical sciences, chemistry, aquaristics, fish farming, aquaculture.
  • Waterproof
  • High accuracy (±0,1 °C ±1,0 digit)
  • Automatic „HOLD“ freezing of constant measuring value
PT1000 probe with PTFE probe and cable.
For use with high temperatures up to 250 °C. The device comes with IP65 /IP67 protection class and a battery lifetime of more than 6 000 hours.
Dodatkowe informacje: Supplied with battery and calibration protocol.