Drukuj

Anti-Phosphothreonine Rabbit Polyclonal Antibody

Dostawca: United States Biological
USBIP4077-11CEA 2810 PLN
USBIP4077-11C
Anti-Phosphothreonine Rabbit Polyclonal Antibody
Przeciwciała
Recommended dilution: ELISA:0.5 ?g/ml; Western Blot:2?g/ml

Type: Primary
Antigen: Phosphothreonine
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR