Drukuj

Anti-Microcystin LR Mouse Monoclonal Antibody [clone: 6D584]

Dostawca: United States Biological
USBIM3889-25EA 3050 PLN
USBIM3889-25
Anti-Microcystin LR Mouse Monoclonal Antibody [clone: 6D584]
Przeciwciała
Recommended dilution: ELISA: 1:10000-1:100000

Type: Primary
Antigen: Microcystin LR
Clonality: monoclonal
Clone: 6D584
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR