Drukuj

Anti-GFP Rabbit polyclonal antibody

Dostawca: United States Biological
USBIG8965-12CEA 2580 PLN
USBIG8965-12C
Anti-GFP Rabbit polyclonal antibody
Przeciwciała
Recommended dilution: Western Blot: 0.5-2?g/ml

Type: Tag
Antigen: GFP (green fluorescent protein)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR