Drukuj

Anti-NR2F6 Goat Polyclonal Antibody

Dostawca: United States Biological
USBIE3451-01CEA 2030 PLN
USBIE3451-01C
Anti-NR2F6 Goat Polyclonal Antibody
Przeciwciała
Recommended dilution: ELISA: 1:32000

Type: Primary
Antigen: NR2F6 (nuclear receptor subfamily 2, group F, member 6)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Bovine, Dog, Human, Mouse, Pig, Rat
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR