Drukuj
Anti-DMAP1 Rabbit Polyclonal Antibody
Dostawca: United States Biological
USBID8073-02GEA 2200 PLN
USBID8073-02G
Anti-DMAP1 Rabbit Polyclonal Antibody
Przeciwciała
Recommended dilution: Western Blot: 1-4?g/ml; Immunochemistry: 5 ?g/ml
Type: Primary
Antigen: DMAP1 (DNA methyltransferase 1 associated protein 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Bovine, Dog, Human, Mouse, Rat