Drukuj

Anti-Clathrin Mouse monoclonal antibody [clone: 5F178]

Dostawca: United States Biological
USBIC5835-04EA 2710 PLN
USBIC5835-04
Anti-Clathrin Mouse monoclonal antibody [clone: 5F178]
Przeciwciała
Recommended dilution: Immunochemistry: 1:10; Immunoblot: 1:100-1:500

Type: Primary
Antigen: Clathrin
Clonality: Monoclonal
Clone: 5F178
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: Bovine, Human, Pig, Rat
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR