Drukuj

Anti-14-3-3 gamma Mouse Monoclonal Antibody [clone: 9i237]

Dostawca: United States Biological
USBI0014-07GEA 3360 PLN
USBI0014-07G
Anti-14-3-3 gamma Mouse Monoclonal Antibody [clone: 9i237]
Przeciwciała
Recommended dilution: Western Blot: 1:1000

Type: Primary
Antigen: 14-3-3 gamma (14-3-3 gamma)
Clonality: Monoclonal
Clone: 9i237
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Bovine, Chicken, Human, Mouse, Rat, Zebrafish
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR