Drukuj

Anti-Protein G Rabbit Polyclonal Antibody

Dostawca: United States Biological
USBI029871EA 2320 PLN
USBI029871
Anti-Protein G Rabbit Polyclonal Antibody
Przeciwciała
Recommended dilution: Western Blot: 0.5-4?g/ml

Type: Tag
Antigen: Protein G
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Bovine, Goat, Guinea pig, Horse, Human, Mouse, Rabbit, Rat, Sheep
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR