Drukuj

Anti-IgG F(c) Goat Polyclonal Antibody (Peroxidase)

Dostawca: Rockland Immunochemicals
ROCK711-1303EA 2240 PLN
ROCK711-1303
Anti-IgG F(c) Goat Polyclonal Antibody (Peroxidase)
Przeciwciała
This product has been assayed against 1.0 µg of Rabbit IgG in a standard capture ELISA using ABTS (2,2’-azino-bis-(3-ethylbenthiazoline-6-sulfonic acid)) as a substrate for 30 minutes at room temperature.

Recommended Dilutions: ELISA: 1:10000 - 1:50000; Western Blot: 1:1000 - 1:10000; Immunochemistry: 1:500 - 1:2500;
Restore with deionized water (or equivalent); 500 µl
Do not add Sodium Azide!

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope: F(c)
Host: Goat
Isotype: IgG
Reactivity: Rabbit
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR