Drukuj

Anti-IgG (H+L) Goat Polyclonal Antibody

Dostawca: Rockland Immunochemicals
ROCK619-101-002EA 1520 PLN
ROCK619-101-002
Anti-IgG (H+L) Goat Polyclonal Antibody
Przeciwciała
Suitable for immunoprecipitation, immunodiffusion, conjugation and most immunological methods requiring lot-to-lot consistency, high titer and specificity.

Recommended Dilutions: ELISA: 1:20000 - 1:100000; Western Blot: 1:2000 - 1:10000;

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Heavy and Light chain
Host: Goat
Isotype:
Reactivity: Armenian Hamster
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR