Drukuj

Anti-IgG (H+L) Donkey Polyclonal Antibody (TRITC (Tetramethylrhodamine Isothiocyanate))

Dostawca: Rockland Immunochemicals
ROCK605-700-002EA 1170 PLN
ROCK605-700-002
Anti-IgG (H+L) Donkey Polyclonal Antibody (TRITC (Tetramethylrhodamine Isothiocyanate))
Przeciwciała
Suitable for immunomicroscopy and flow cytometry or FACS analysis as well as other antibody based fluorescent assays requiring lot-to-lot consistency.

Recommended Dilutions: FLISA: 1:10000 - 1:50000; Immunofluorescence: 1:1000 - 1:5000; Flow Cytometry: 1:500 - 1:2500
Restore with deionized water (or equivalent); 1.0 ml

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: TRITC (Tetramethylrhodamine Isothiocyanate)
Epitope: Heavy and Light chain
Host: Donkey
Isotype: IgG
Reactivity: Goat
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR