Drukuj

Anti-IgG (H+L) Rabbit Polyclonal Antibody (Texas Red®)

Dostawca: Rockland Immunochemicals
ROCK605-4902EA 1390 PLN
ROCK605-4902
Anti-IgG (H+L) Rabbit Polyclonal Antibody (Texas Red®)
Przeciwciała
Secondary Rabbit Anti-IgG (H&L) Reacts with Goat

Recommended Dilutions: FLISA: 1:10000 - 1:50000; Immunofluorescence: 1:1000 - 1:5000;
Restore with deionized water (or equivalent); 1.0 ml

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Texas Red®
Epitope: Heavy and Light chain
Host: Rabbit
Isotype: IgG
Reactivity: Goat
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR